missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N,N'-Methylenebisacrylamide, 99+%, Electrophoresis Grade
Click to view available options
Quantity:
25 g
100 g
250 g
Description
N,N'-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N,N′-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.
Solubility
Soluble in water or methanol
Notes
Air & Light Sensitive. Store in the dark. Store under dry inert gas. Incompatible with light, heat, air and oxidizing agents. Store at 4°C.
N,N′-Methylenebisacrylamide is used in the preparation of polyacrylamide gels for electrophoretic separation of nucleic acids and proteins. It has also been used as a test compound for studying the biochemical isolation of insoluble Tau in transgenic mouse models of tauopathy by examining the two main isolation methods sarkosyl and formic acid extraction.
Solubility
Soluble in water or methanol
Notes
Air & Light Sensitive. Store in the dark. Store under dry inert gas. Incompatible with light, heat, air and oxidizing agents. Store at 4°C.
Specifications
Melting Point | >300°C |
Color | White |
Density | 1.235 g/mL |
Quantity | 25 g |
Beilstein | 1706297 |
Sensitivity | Air and light sensitive |
Solubility Information | Soluble in water or methanol |
Formula Weight | 154.17 |
Percent Purity | ≥99% |
Grade | Electrophoresis |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'