Learn More
DMF, Dimethylformamide, Sequencing, Fisher BioReagents
Catalog Number p-25570
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Glass Bottle
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
68-12-2
C3H7NO
73.10
MFCD00003284
ZMXDDKWLCZADIW-UHFFFAOYSA-N
dimethylformamide, dimethyl formamide, n,n-dimethylmethanamide, n-formyldimethylamine, formamide, n,n-dimethyl, dmf, dimethylformamid, dimetilformamide, dwumetyloformamid, dmfa
6228
CHEBI:17741
N,N-dimethylformamide
CN(C)C=O
  • DMF for sequencing and peptide synthesis
  • Useful for protein chemistry due to low amine content
  • DMF is a commonly used solvent in solid phase peptide synthesis
  • Intended for research use only
TRUSTED_SUSTAINABILITY
pH 6 to 8
Melting Point -61°C
Absorbance Pass Test, 1 max. at 270nm, 0.3 max. at 275nm, 0.1 max. at 295nm, 0.05 max. at 310nm, 0.01 max. (340nm to 400nm) at 340nm
Color Colorless
Assay Percent Range ≥99.5 %
Linear Formula HCONCH3CH3
UN Number 2265
Merck Index 15, 3267
Formula Weight 73.09
Percent Purity ≥99.5%
Grade Sequencing
Decomposition Information >350°C
Density 0.945g/cm³
Boiling Point 153°C
Quantity 500 mL
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.005% max.
Titratable Acid 0.0005mEq/g max.
Titratable Base 0.003mEq/g max.
Vapor Pressure 4.9mbar at 20°C
Viscosity 0.8 mPa/s at 20°C
Water 0.03% max.
Chemical Name or Material N, N-Dimethylformamide
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N,N-Dimethylformamide
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Reproductive toxicity Category 1B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H312-Harmful in contact with skin.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.