Learn More
N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride, Mikromol
Specifications
Specifications
| Chemical Name or Material | N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride |
| CAS | 63434-11-7 |
| Analyte or Component Names | N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C25 H38 N2 O4 . 2 Cl H |
| Certifications/Compliance | ISO 9001 |
| Synonym | Verapamil Hydrochloride Imp. A (EP), Verapamil Imp. A (EP), N,N'-Bis[2-(3,4-dimethoxyphenyl)ethyl]-N,N'-dimethylpropane-1,3-diamine Dihydrochloride, Verapamil Hydrochloride Imp. A (EP) as Dihydrochloride, Verapamil Hydrochloride Impurity A as Dihydrochloride, Verapamil Impurity A as Dihydrochloride |
| Solution Type | Neat |
| SMILES | Cl.Cl.COc1ccc(CCN(C)CCCN(C)CCc2ccc(OC)c(OC)c2)cc1OC |
| Molecular Weight (g/mol) | 503.51 |
| Show More |