Learn More
N,N,2-Trimethyl-3,3-diphenylpropan-1-amine Hydrochloride, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
N,N,2-Trimethyl-3,3-diphenylpropan-1-amine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | N,N,2-Trimethyl-3,3-diphenylpropan-1-amine Hydrochloride |
| CAS | 63905-14-6 |
| Analyte or Component Names | N,N,2-Trimethyl-3,3-diphenylpropan-1-amine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H23 N . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Propylamine, N,N,2-trimethyl-3,3-diphenyl-, hydrochloride (7CI), Benzenepropanamine, N,N,beta-trimethyl-gamma-phenyl-, hydrochloride (1:1), Benzenepropanamine, N,N,beta-trimethyl-gamma-phenyl-, hydrochloride (9CI) |
| InChI Formula | InChI=1S/C18H23N.ClH/c1-15(14-19(2)3)18(16-10-6-4-7-11-16)17-12-8-5-9-13-17;/h4-13,15,18H,14H2,1-3H3;1H |
| SMILES | Cl.CC(CN(C)C)C(c1ccccc1)c2ccccc2 |
| IUPAC Name | N,N,2-trimethyl-3,3-diphenylpropan-1-amine;hydrochloride |
| Show More |