missing translation for 'onlineSavingsMsg'
Learn More

N-Methylcarbazole, 99%, Thermo Scientific Chemicals

Catalog Number p-4259014
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
1484-12-4
C13H11N
181.24
SDFLTYHTFPTIGX-UHFFFAOYSA-N
n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole
15142
9-methylcarbazole
CN1C2=CC=CC=C2C3=CC=CC=C31
This item is not returnable. View return policy
1484-12-4
C13H11N
181.24
SDFLTYHTFPTIGX-UHFFFAOYSA-N
n-methylcarbazole, 9-methyl-9h-carbazole, 9h-carbazole, 9-methyl, carbazole, 9-methyl, n-methyldibenzopyrrole, unii-76sop090pg, ccris 6846, n-methyl carbazole, pubchem17421, 9-methylcarbazole
15142
9-methylcarbazole
CN1C2=CC=CC=C2C3=CC=CC=C31

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 86.0°C to 89.0°C
Color White
Boiling Point 163.0°C to 167.0°C (0.4 Torr)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 181.24
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material N-Methylcarbazole
Show More Show Less
compliance-icons
Product Identifier
  • N-Methylcarbazole
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'