missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Methyl-L-proline, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.
Solubility
Soluble in water and methanol(both 50 mg/ml-clear-colorless solution)
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
N-Methyl-L-proline, is an aminoacid which is widely used in pharmaceuticals and food industry.
Solubility
Soluble in water and methanol(both 50 mg/ml-clear-colorless solution)
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Color | White |
Refractive Index | 1.5 |
Solubility Information | Soluble in water and methanol(both 50mg/ml-clear-colorless solution) |
Formula Weight | 129.16 |
Percent Purity | 98% |
Quantity | 1 g |
Physical Form | Powder |
Chemical Name or Material | N-Methyl-L-proline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'