missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(6-phenylpyrid-3-yl)methylamine, 97%, Thermo Scientific™

Catalog Number p-3527266
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
879896-40-9
C13H14N2
198.27
MFCD09064942
OLZSHISXLYLLBI-UHFFFAOYSA-N
n-methyl 6-phenylpyrid-3-yl methylamine, methyl 6-phenylpyridin-3-yl methyl amine, 3-pyridinemethanamine,n-methyl-6-phenyl, methyl 6-phenyl 3-pyridyl methyl amine, n-methyl-1-6-phenylpyridin-3-yl methanamine
24229465
N-methyl-1-(6-phenylpyridin-3-yl)methanamine
CNCC1=CN=C(C=C1)C1=CC=CC=C1
This item is not returnable. View return policy
879896-40-9
C13H14N2
198.27
MFCD09064942
OLZSHISXLYLLBI-UHFFFAOYSA-N
n-methyl 6-phenylpyrid-3-yl methylamine, methyl 6-phenylpyridin-3-yl methyl amine, 3-pyridinemethanamine,n-methyl-6-phenyl, methyl 6-phenyl 3-pyridyl methyl amine, n-methyl-1-6-phenylpyridin-3-yl methanamine
24229465
N-methyl-1-(6-phenylpyridin-3-yl)methanamine
CNCC1=CN=C(C=C1)C1=CC=CC=C1
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 198.27
Percent Purity 97%
Chemical Name or Material N-methyl(6-phenylpyrid-3-yl)methylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'