Learn More
N-Methyl-4-(phenoxymethyl)benzylamine, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | N-methyl-1-[4-(phenoxymethyl)phenyl]methanamine |
Formula Weight | 227.31 |
Percent Purity | 95% |
Chemical Name or Material | N-methyl-4-(phenoxymethyl)benzylamine |