missing translation for 'onlineSavingsMsg'
Learn More

n-Heptane, 99%, for biochemistry, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4813577
Click to view available options
Quantity:
1 L
2.5 L
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
142-82-5
C7H16
100.21
MFCD00009544
IMNFDUFMRHMDMM-UHFFFAOYSA-N
n-heptane, dipropylmethane, heptan, heptyl hydride, dipropyl methane, gettysolve-c, skellysolve c, heptanen, eptani, heptan polish
8900
CHEBI:43098
heptane
CCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point -91.0°C
Density 0.6830g/mL
Boiling Point 98.0°C
Flash Point −4°C
Assay Percent Range 98.5% min. (GC)
Linear Formula CH3(CH2)5CH3
Refractive Index 1.3850 to 1.3890
Quantity 1 L
Merck Index 15,4694
Specific Gravity 0.683
Solubility Information Solubility in water: practically insoluble. Other solubilities: soluble in alcohol, ether and chloroform
Viscosity 0.4 mPa.s (20°C)
Formula Weight 100.2
Percent Purity 99%
Grade Biochemistry
Chemical Name or Material n-Heptane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'