missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-S-tert-butyl-L-cysteine, 98%, Thermo Scientific™

Catalog Number p-7039886
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
67436-13-9
C22H25NO4S
399.505
MFCD00037130
IXAYZHCPEYTWHW-IBGZPJMESA-N
fmoc-cys tbu-oh, fmoc-s-tert-butyl-l-cysteine, fmoc-s-t-butyl-l-cysteine, n-fmoc-s-tert-butyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butylthio propanoic acid, fmoc-r-2-amino-3-s-t-butylthio butanoic acid, 2r-3-tert-butylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1716, pubchem19015, fmoc-cys but-oh
7009544
(2R)-3-tert-butylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
CC(C)(C)SCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
This item is not returnable. View return policy
67436-13-9
C22H25NO4S
399.505
MFCD00037130
IXAYZHCPEYTWHW-IBGZPJMESA-N
fmoc-cys tbu-oh, fmoc-s-tert-butyl-l-cysteine, fmoc-s-t-butyl-l-cysteine, n-fmoc-s-tert-butyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-tert-butylthio propanoic acid, fmoc-r-2-amino-3-s-t-butylthio butanoic acid, 2r-3-tert-butylsulfanyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1716, pubchem19015, fmoc-cys but-oh
7009544
(2R)-3-tert-butylsulfanyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
CC(C)(C)SCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Beilstein 4577031
Optical Rotation +24° (c=1 in DMF)
Formula Weight 399.5
Percent Purity 98%
Chemical Name or Material N-Fmoc-S-tert-butyl-L-cysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'