missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-S-(4-methoxybenzyl)-L-cysteine, 95%, Thermo Scientific™

Catalog Number p-7054663
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
141892-41-3
C26H25NO5S
463.548
MFCD00065635
IWZGYHFOLFRYPK-DEOSSOPVSA-N
fmoc-cys pmeobzl-oh, fmoc-s-p-methoxybenzyl-l-cysteine, fmoc-cys mob-oh, n-fmoc-s-4-methoxybenzyl-l-cysteine, fmoc-s-4-methoxybenzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-s-4-methoxybenzyl-l-cysteine, fmoc-cys 4-meobzl-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, pubchem19018
11236605
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
COC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24
This item is not returnable. View return policy
141892-41-3
C26H25NO5S
463.548
MFCD00065635
IWZGYHFOLFRYPK-DEOSSOPVSA-N
fmoc-cys pmeobzl-oh, fmoc-s-p-methoxybenzyl-l-cysteine, fmoc-cys mob-oh, n-fmoc-s-4-methoxybenzyl-l-cysteine, fmoc-s-4-methoxybenzyl-l-cysteine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-methoxybenzyl thio propanoic acid, n-alpha-9-fluorenylmethyloxycarbonyl-s-4-methoxybenzyl-l-cysteine, fmoc-cys 4-meobzl-oh, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-methoxyphenyl methyl sulfanyl propanoic acid, pubchem19018
11236605
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(4-methoxyphenyl)methylsulfanyl]propanoic acid
COC1=CC=C(C=C1)CSCC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Formula Weight 463.55
Percent Purity 95%
Quantity 5 g
Physical Form Powder
Chemical Name or Material N-Fmoc-S-(4-methoxybenzyl)-L-cysteine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'