missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog Number 11389729
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
86123-10-6
C24H21NO4
387.44
MFCD00062955
SJVFAHZPLIXNDH-JOCHJYFZSA-N
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
978332
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
This item is not returnable. View return policy
86123-10-6
C24H21NO4
387.44
MFCD00062955
SJVFAHZPLIXNDH-JOCHJYFZSA-N
fmoc-d-phenylalanine, fmoc-d-phe-oh, n-fmoc-d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-d-phenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-phenylpropanoic acid, fmoc-r-beta-phenylalanine, n-9-fluorenylmethoxycarbonyl-d-phenylalanine, d-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-phenylalanine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-phenyl-propanoic acid
978332
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-phenylpropanoic acid
OC(=O)[C@@H](CC1=CC=CC=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 181°C to 185°C
Quantity 25 g
Optical Rotation +40° (c=1 in DMF)
Formula Weight 387.44
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'