missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-chloro-L-phenylalanine, 98%, Thermo Scientific Chemicals

Catalog Number 15430149
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
175453-08-4
C24H20ClNO4
421.877
MFCD00080281
CQPNKLNINBUUOM-QFIPXVFZSA-N
fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
2734471
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O
This item is not returnable. View return policy
175453-08-4
C24H20ClNO4
421.877
MFCD00080281
CQPNKLNINBUUOM-QFIPXVFZSA-N
fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
2734471
(2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Beilstein 9437838
Formula Weight 421.87
Percent Purity 98%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material N-Fmoc-4-chloro-L-phenylalanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'