missing translation for 'onlineSavingsMsg'
Learn More

n-Docosane, 99%, Thermo Scientific Chemicals

Catalog Number p-3778485
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-97-0
C22H46
310.61
MFCD00009348
HOWGUJZVBDQJKV-UHFFFAOYSA-N
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
12405
CHEBI:46050
docosane
CCCCCCCCCCCCCCCCCCCCCC
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
629-97-0
C22H46
310.61
MFCD00009348
HOWGUJZVBDQJKV-UHFFFAOYSA-N
n-docosane, dokosan, unii-ow99q363ko, heneicosane, methyl, ch3-ch2 20-ch3, docosane, analytical standard, twt, normal-docosane, docosane, n, docosane
12405
CHEBI:46050
docosane
CCCCCCCCCCCCCCCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 43.0°C to 46.0°C
Color White
Density 0.7700g/mL
Boiling Point 369.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)20CH3
Quantity 25 g
Beilstein 01, 174
Specific Gravity 0.77
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene and toluene
Formula Weight 310.6
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material n-Docosane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'