missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltriethoxysilane, 98%, Thermo Scientific Chemicals

Catalog Number p-7046960
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
2943-73-9
C16H36O3Si
304.546
MFCD00069132
BAAAEEDPKUHLID-UHFFFAOYSA-N
n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy
76261
decyl(triethoxy)silane
CCCCCCCCCC[Si](OCC)(OCC)OCC
This item is not returnable. View return policy
2943-73-9
C16H36O3Si
304.546
MFCD00069132
BAAAEEDPKUHLID-UHFFFAOYSA-N
n-decyltriethoxysilane, decyl triethoxy silane, silane, decyltriethoxy
76261
decyl(triethoxy)silane
CCCCCCCCCC[Si](OCC)(OCC)OCC

n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltriethoxysilane is used as coupling agent and as a chemical additive. It is also used as an intermediate of OLED, pharmaceuticals and in cosmetics. It is employed in many industries like adhesives, coatings, crude oil, glass fibers, filler treatment, polymer modification, printing, elastomers, sealants, textiles and thermoplastics.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Boiling Point 150°C (8mmHg)
Quantity 25 g
Linear Formula CH3(CH2)9Si(OCH2CH3)3
Beilstein 2413155
Flash Point >110°C (230°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 304.55
Percent Purity 98%
Refractive Index 1.422
Sensitivity Moisture sensitive
Density 0.879
Chemical Name or Material n-Decyltriethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'