missing translation for 'onlineSavingsMsg'
Learn More

n-Butyldimethylchlorosilane, 97+%, Thermo Scientific™

Catalog Number p-4228480
Click to view available options
Quantity:
10 mL
50 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
1000-50-6
C6H15ClSi
150.72
MFCD00015727
MXOSTENCGSDMRE-UHFFFAOYSA-N
butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
66079
butyl-chloro-dimethylsilane
CCCC[Si](C)(C)Cl
This item is not returnable. View return policy
1000-50-6
C6H15ClSi
150.72
MFCD00015727
MXOSTENCGSDMRE-UHFFFAOYSA-N
butylchlorodimethylsilane, butyldimethylchlorosilane, n-butyldimethylchlorosilane, butyldimethylsilyl chloride, 1-butyldimethylchlorosilane, silane, butylchlorodimethyl, butyl chloro dimethylsilane, n-butyl-dimethylchlorosilane, butyl-chloro-dimethyl-silane, n-butyldimethylsilyl chloride
66079
butyl-chloro-dimethylsilane
CCCC[Si](C)(C)Cl
CAS Min % 97.0
CAS Max % 100.0
Color Colorless
Density 0.8750g/mL
Boiling Point 138.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Linear Formula Cl(CH3)2Si(CH2)3CH3
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Quantity 10 mL
Specific Gravity 0.875
Solubility Information Solubility in water: insoluble,reacts
Formula Weight 150.72
Percent Purity 97+%
Physical Form Liquid
Chemical Name or Material n-Butyldimethylchlorosilane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • n-Butyldimethylchlorosilane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'