Learn More
n-Butylbenzene-d14, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | Butylbenzene-D14 |
| CAS | 634897-78-2 |
| Molecular Formula | C10 D14 |
| Quantity | 0.25 g |
| Synonym | Benzene-1,2,3,4,5-d5, 6-(butyl-1,1,2,2,3,3,4,4,4-d9)- (ACI), 6-(Butyl-1,1,2,2,3,3,4,4,4-d9)benzene-1,2,3,4,5-d5 (ACI), Benzene-d5, butyl-d9- (9 CI), Butyl-d9-benzene-d5, Butylbenzene-d14, Butylbenzene-D14 |
| InChI Formula | InChI=1 S/C10H14/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9 H,2-3,7H2,1H3/i1D3,2D2,3D2,4 D,5 D,6 D,7D2,8 D,9 D |
| SMILES | [2 H]c1c([2 H])c([2 H])c(c([2 H])c1[2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])[2 H] |
| IUPAC Name | 1,2,3,4,5-pentadeuterio-6-(1,1,2,2,3,3,4,4,4-nonadeuteriobutyl)benzene |
| Molecular Weight (g/mol) | 148.304 |
| Formula Weight | 148.197 g/mol |
| Show More |