Learn More
N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™
Catalog Number p-7072877
Click to view available options
Quantity:
500 mL
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
Metal can
123-86-4
116.16
9445
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
CCCCOC(C)=O
123-86-4
116.16
9445
DKPFZGUDAPQIHT-UHFFFAOYSA-N
butyl acetate
CCCCOC(C)=O
N-Butyl Acetate, Extra Pure, SLR
TRUSTED_SUSTAINABILITY
Melting Point -90°C
Color Colorless
pH 6.2
Boiling Point 126°C
Packaging Amber glass bottle
Molecular Formula C6H12O2
Quantity 500 mL
Vapor Density 4
Formula Weight 116.16g/mol
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • n-Butyl acetate
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.