Learn More
N-Butyl Acetate, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 mL
2.5 L
5 L
10 L
25 L
Packaging:
Amber glass bottle
Metal can
Description
N-Butyl Acetate, Extra Pure, SLR
Specifications
Melting Point | -90°C |
Color | Colorless |
pH | 6.2 |
Boiling Point | 126°C |
Packaging | Metal can |
Molecular Formula | C6H12O2 |
Quantity | 10 L |
Vapor Density | 4 |
Formula Weight | 116.16g/mol |
Physical Form | Liquid |