Learn More
n-Butyl-3,3,4,4,4-d5 Alcohol, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1-Butanol-3,3,4,4,4-D5 |
| CAS | 64118-16-7 |
| Molecular Formula | C4 D5 H5 O |
| Quantity | 1 g |
| Synonym | 1-Butan-3,3,4,4,4-d5-ol (9 CI), Butan-3,3,4,4,4-d5-1-ol, Butyl-3,3,4,4,4-d5 alcohol, n-Butyl-3,3,4,4,4-d5 alcohol, 1-Butanol-3,3,4,4,4-D5 |
| InChI Formula | InChI=1 S/C4H10O/c1-2-3-4-5/h5H,2-4H2,1H3/i1D3,2D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])CCO |
| IUPAC Name | 3,3,4,4,4-pentadeuteriobutan-1-ol |
| Molecular Weight (g/mol) | 79.152 |
| Formula Weight | 79.105 g/mol |
| Show More |