missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-Boc-O-benzyl-D-tyrosine, 95%

Brand:  Alfa Aesar™ H63427.03

Additional Details : CAS Number : 63769-58-4 Weight : 0.00100kg

Quantity 1g

 View more versions of this product

Product Code. 15430119

  • 423.82 NOK / 1g


Estimated Shipment
09-09-2020
Add to basket

Description

Description

Applications
N-Boc-O-benzyl-D-tyrosine is used as a intermediate for pharmaceutical and chemical research.

Solubility
Slightly soluble in water.

Notes
Store in freezer at(-20°C). Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

63769-58-4
371.433
ZAVSPTOJKOFMTA-GOSISDBHSA-N
6992575
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
C21H25NO5
MFCD00038249
boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid

Specifications

N-Boc-O-benzyl-D-tyrosine
C21H25NO5
boc-d-tyr bzl-oh, boc-o-benzyl-d-tyrosine, n-boc-o-benzyl-d-tyrosine, boc-o-benzyl-d-tyr, r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-4-benzyloxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, ambotzbaa1381, boc-dtyr bn-oh, pubchem14949, boc-d-tyr bzl oh
ZAVSPTOJKOFMTA-GOSISDBHSA-N
(2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-(4-phenylmethoxyphenyl)propanoic acid
6992575
Solid
1g
63769-58-4
MFCD00038249
Slightly soluble in water.
CC(C)(C)OC(=O)NC(CC1=CC=C(C=C1)OCC2=CC=CC=C2)C(=O)O
371.433
371.43
95%
Videos
SDS Documents