missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-L-tryptophanol, 98%, Thermo Scientific™

Catalog Number p-4090062
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
82689-19-8
C16H22N2O3
290.36
MFCD00235953
JEFQUFUAEKORKL-LBPRGKRZSA-N
n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
7019533
tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
This item is not returnable. View return policy
82689-19-8
C16H22N2O3
290.36
MFCD00235953
JEFQUFUAEKORKL-LBPRGKRZSA-N
n-boc-l-tryptophanol, boc-tryptophanol, boc-l-tryptophanol, boc-trp-ol, s-tert-butyl 1-hydroxy-3-1h-indol-3-yl propan-2-yl carbamate, s-3-2-boc-amino-3-hydroxypropyl-indole, boc-d-trp-ol, n-alpha-boc-l-tryptophanol, boc-trypotophanol, boc-l-trp-ol
7019533
tert-butyl N-[(2S)-1-hydroxy-3-(1H-indol-3-yl)propan-2-yl]carbamate
CC(C)(C)OC(=O)N[C@H](CO)CC1=CNC2=CC=CC=C12
CAS Min % 97.5
CAS Max % 100.0
Melting Point 118°C to 122°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Specific Rotation -28° to -32° (20°C, 589nm) (C=1, Methanol)
Formula Weight 290.36
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-BOC-L-tryptophanol
Show More Show Less
compliance-icons
Product Identifier
  • N-alpha-BOC-L-tryptophanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'