Learn More
N-Boc-L-prolinal, 96%, Thermo Scientific Chemicals
Catalog Number p-7029063
1350.06 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
69610-41-9
C10H17NO3
199.25
MFCD00274186,MFCD00801206
YDBPZCVWPFMBDH-QMMMGPOBSA-N
n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
2734674
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC[C@H]1C=O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
69610-41-9
C10H17NO3
199.25
MFCD00274186,MFCD00801206
YDBPZCVWPFMBDH-QMMMGPOBSA-N
n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal
2734674
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC[C@H]1C=O
Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.
Solubility
Slightly soluble water. Soluble in chloroform.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
TRUSTED_SUSTAINABILITY
Density 1.06
Boiling Point 228°C
Flash Point >110°C (230°F)
Refractive Index 1.4589
Quantity 1 g
Sensitivity Air sensitive
Solubility Information Slightly soluble water. Soluble in chloroform.
Optical Rotation −95° (c=1 in Chloroform)
Formula Weight 199.25
Percent Purity 96%
Chemical Name or Material N-Boc-L-prolinal