missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-L-beta-proline, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (slightly).
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
N-Boc-L-beta-proline is an important raw material and intermediate used in pharmaceuticals, agrochemicals and dyestuff.
Solubility
Soluble in water (slightly).
Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in water (slightly). |
Optical Rotation | +15° (c=0.8 in Chloroform) |
Formula Weight | 215.25 |
Percent Purity | 95% |
Chemical Name or Material | N-Boc-L-beta-proline |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'