missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-beta-prolinol, 97+%, Thermo Scientific™

Catalog Number p-7038295
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
138108-72-2
C10H19NO3
201.266
MFCD03094725
HKIGXXRMJFUUKV-MRVPVSSYSA-N
r-tert-butyl 3-hydroxymethyl pyrrolidine-1-carboxylate, r-1-boc-3-hydroxymethyl pyrrolidine, r-1-boc-3-hydroxymethylpyrrolidine, tert-butyl 3r-3-hydroxymethyl pyrrolidine-1-carboxylate, r-3-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-1-boc-3-hydroxymethyl-pyrrolidine, n-boc-d-beta-prolinol, r-boc-3-pyrrolidinemethanol, r-n-boc-pyrrolidine-3-methanol, r-3-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester
1514340
tert-butyl (3R)-3-(hydroxymethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)CO
This item is not returnable. View return policy
138108-72-2
C10H19NO3
201.266
MFCD03094725
HKIGXXRMJFUUKV-MRVPVSSYSA-N
r-tert-butyl 3-hydroxymethyl pyrrolidine-1-carboxylate, r-1-boc-3-hydroxymethyl pyrrolidine, r-1-boc-3-hydroxymethylpyrrolidine, tert-butyl 3r-3-hydroxymethyl pyrrolidine-1-carboxylate, r-3-hydroxymethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-1-boc-3-hydroxymethyl-pyrrolidine, n-boc-d-beta-prolinol, r-boc-3-pyrrolidinemethanol, r-n-boc-pyrrolidine-3-methanol, r-3-hydroxymethylpyrrolidine-1-carboxylic acid tert-butyl ester
1514340
tert-butyl (3R)-3-(hydroxymethyl)pyrrolidine-1-carboxylate
CC(C)(C)OC(=O)N1CCC(C1)CO

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water

Notes
Store in a cool ventilated place. Avoid strong oxidizing materials.
TRUSTED_SUSTAINABILITY
Density 1.089
Quantity 250 mg
Solubility Information Slightly soluble in water
Formula Weight 201.27
Percent Purity ≥97%
Chemical Name or Material N-Boc-D-beta-prolinol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'