missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-D-alaninol, 98%, ee 98%, Thermo Scientific Chemicals

Catalog Number p-7030318
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
106391-86-0
C8H17NO3
175.23
MFCD00235912
PDAFIZPRSXHMCO-ZCFIWIBFSA-N
n-boc-d-alaninol, boc-d-alaninol, r-2-boc-amino-1-propanol, tert-butyl n-2r-1-hydroxypropan-2-yl carbamate, r-tert-butyl 1-hydroxypropan-2-yl carbamate, n-alpha-t-boc-d-alaninol, r-tert-butyl 1-hydroxypropan-2-ylcarbamate, r-+-2-tert-butoxycarbonylamino-1-propanol, tert-butyl 2r-1-hydroxypropan-2-yl carbamate, tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate
13200309
tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
C[C@H](CO)NC(=O)OC(C)(C)C
This item is not returnable. View return policy
106391-86-0
C8H17NO3
175.23
MFCD00235912
PDAFIZPRSXHMCO-ZCFIWIBFSA-N
n-boc-d-alaninol, boc-d-alaninol, r-2-boc-amino-1-propanol, tert-butyl n-2r-1-hydroxypropan-2-yl carbamate, r-tert-butyl 1-hydroxypropan-2-yl carbamate, n-alpha-t-boc-d-alaninol, r-tert-butyl 1-hydroxypropan-2-ylcarbamate, r-+-2-tert-butoxycarbonylamino-1-propanol, tert-butyl 2r-1-hydroxypropan-2-yl carbamate, tert-butyl n-1r-2-hydroxy-1-methyl-ethyl carbamate
13200309
tert-butyl N-[(2R)-1-hydroxypropan-2-yl]carbamate
C[C@H](CO)NC(=O)OC(C)(C)C

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 58°C to 62°C
Formula Weight 175.23
Percent Purity 98%
Quantity 1 g
Optical Rotation +11° (c=1 in Chloroform)
Assay ee 98%
Chemical Name or Material N-Boc-D-alaninol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'