missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3-(2-naphthyl)-L-alanine, 97%, Thermo Scientific Chemicals

Catalog Number p-7054791
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
58438-04-3
C18H21NO4
315.369
MFCD00079671
URKWHOVNPHQQTM-HNNXBMFYSA-N
boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
7020908
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O
This item is not returnable. View return policy
58438-04-3
C18H21NO4
315.369
MFCD00079671
URKWHOVNPHQQTM-HNNXBMFYSA-N
boc-3-2-naphthyl-l-alanine, boc-2-nal-oh, boc-l-2-naphthylalanine, n-tert-butoxycarbonyl-3-2-naphthyl-l-alanine, boc-l-3-2-naphthyl-alanine, s-boc-3-2-naphthyl-l-alanine, s-n-boc-2-naphthylalanine, boc-2-l-naphthylalanine, n-boc-3-2-naphthyl-l-alanine, s-2-tert-butoxycarbonyl amino-3-naphthalen-2-yl propanoic acid
7020908
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-naphthalen-2-ylpropanoic acid
CC(C)(C)OC(=O)NC(CC1=CC2=CC=CC=C2C=C1)C(=O)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point ∼95°C
Color White
Beilstein 4326467
Formula Weight 315.37
Percent Purity 97%
Quantity 1 g
Physical Form Solid
Chemical Name or Material N-Boc-3-(2-naphthyl)-L-alanine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'