missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-2-methyl-L-serine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
Description
N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.
Solubility
Soluble in dimethylformamide.
Notes
Incompatible with reducing agents, water and strong oxidizing agents.
N-Boc-2-methyl-L-serine is used as a cocatalyst in the Diels-Alder reactions between cyclopentadiene and methyl acrylate in the presence of Lewis acids.
Solubility
Soluble in dimethylformamide.
Notes
Incompatible with reducing agents, water and strong oxidizing agents.
Specifications
Quantity | 250 mg |
Solubility Information | Soluble in dimethylformamide. |
Formula Weight | 219.24 |
Percent Purity | 97% |
Chemical Name or Material | N-Boc-2-methyl-L-serine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'