missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-2-bromobenzylamine, 97%, Thermo Scientific™

Catalog Number p-7052659
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
162356-90-3
C12H16BrNO2
286.169
MFCD05664385
DFNZFCPEUDSNEO-UHFFFAOYSA-N
tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
7018300
tert-butyl N-[(2-bromophenyl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br
This item is not returnable. View return policy
162356-90-3
C12H16BrNO2
286.169
MFCD05664385
DFNZFCPEUDSNEO-UHFFFAOYSA-N
tert-butyl 2-bromobenzylcarbamate, n-boc-2-bromobenzylamine, n-tert-butoxycarbonyl-2-bromobenzylamine, tert-butyl n-2-bromophenyl methyl carbamate, 2-bromo-benzyl-carbamic acid tert-butyl ester, tert-butyl 2-bromobenzyl carbamate, 2-bromobenzyl carbamic acid tert-butyl ester, 1-tert-butoxycarbonylaminomethyl-2-bromobenzene, carbamic acid, 2-bromophenyl methyl-, 1,1-dimethylethyl ester, pubchem7804
7018300
tert-butyl N-[(2-bromophenyl)methyl]carbamate
CC(C)(C)OC(=O)NCC1=CC=CC=C1Br

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 50°C to 54°C
Color White to Brown
Flash Point >110°C (230°F)
Quantity 250 mg
UN Number UN3077
Formula Weight 286.17
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Boc-2-bromobenzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'