missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Boc-1-Boc-L-tryptophan, 95%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Soluble in dimethylformamide (1 mmole in 2 ml).
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
N-Boc-1-Boc-L-tryptophan is generally used as a intermediate in pharmaceutical and chemical research.
Solubility
Soluble in dimethylformamide (1 mmole in 2 ml).
Notes
Store at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications
Quantity | 1 g |
Solubility Information | Soluble in dimethylformamide (1 mmole in 2 ml). |
Formula Weight | 404.46 |
Percent Purity | 95% |
Chemical Name or Material | N-Boc-1-Boc-L-tryptophan |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'