missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylquininium chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-7035700
523.22 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67174-25-8
C27H31ClN2O2
451.007
MFCD00198105
JYDIJFKNXHPWBJ-PRJMIWRBSA-M
n-benzylquininiumchloride chiralphase-transfercatalyst
133611806
(R)-[(2S,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
67174-25-8
C27H31ClN2O2
451.007
MFCD00198105
JYDIJFKNXHPWBJ-PRJMIWRBSA-M
n-benzylquininiumchloride chiralphase-transfercatalyst
133611806
(R)-[(2S,4R,5S)-1-benzyl-5-ethenyl-1-azoniabicyclo[2.2.2]octan-2-yl]-(6-methoxyquinolin-4-yl)methanol;chloride
COC1=CC2=C(C=CN=C2C=C1)C(C3CC4CC[N+]3(CC4C=C)CC5=CC=CC=C5)O.[Cl-]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 200°C to 205°C (decomposition)
Quantity 1 g
Optical Rotation −235° (c=1.5 in Water)
Formula Weight 451.01
Percent Purity 95%
Chemical Name or Material N-Benzylquininium chloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'