missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Benzylethanolamine, 96%, Thermo Scientific Chemicals
837.64 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
500 g
2500 g
Description
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.
Solubility
Soluble in water 162 g/L (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents..
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.
Solubility
Soluble in water 162 g/L (25°C).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents..
Specifications
Melting Point | -30°C |
Density | 1.063 |
pH | 11 |
Boiling Point | 240°C |
Flash Point | 130°C (266°F) |
Refractive Index | 1.543 |
Quantity | 500 g |
Beilstein | 775164 |
Solubility Information | Soluble in water 162g/L (25°C). |
Formula Weight | 151.21 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'