missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-2-chloro-N-isopropylbenzamide, 97%, Thermo Scientific™

Catalog Number p-7040032
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
55578-23-9
C17H18ClNO
287.787
MFCD00783909
ZNMYNGJECVXOIL-UHFFFAOYSA-N
n-benzyl-2-chloro-n-isopropylbenzamide
5192029
N-benzyl-2-chloro-N-propan-2-ylbenzamide
CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=CC=C2Cl
This item is not returnable. View return policy
55578-23-9
C17H18ClNO
287.787
MFCD00783909
ZNMYNGJECVXOIL-UHFFFAOYSA-N
n-benzyl-2-chloro-n-isopropylbenzamide
5192029
N-benzyl-2-chloro-N-propan-2-ylbenzamide
CC(C)N(CC1=CC=CC=C1)C(=O)C2=CC=CC=C2Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 287.79
Percent Purity 97%
Chemical Name or Material N-Benzyl-2-chloro-N-isopropylbenzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'