missing translation for 'onlineSavingsMsg'
Learn More

N-(3-Hydroxyphenyl)thiourea, 97%, Thermo Scientific Chemicals

Catalog Number p-7045253
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3394-05-6
MFCD00022167
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
3394-05-6
MFCD00022167

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 175°C to 180°C
Odor Odorless
Molecular Formula C8H8N2O6
Quantity 2 g
Beilstein 2090372
Synonym 4,5-dinitro veratrole, 1,2-dimethoxy-4,5-dinitro-benzene, 4,5-dimethoxy-1,2-dinitrobenzene, 4,5-dinitroveratrole, acmc-1ckho, ghl.pd_mitscher_leg0.933, benzene,1,2-dimethoxy-4,5-dinitro, benzene, 1,2-dimethoxy-4,5-dinitro
InChI Key WFDHPWTYKOAFBJ-UHFFFAOYSA-N
SMILES COC1=C(C=C(C(=C1)[N+](=O)[O-])[N+](=O)[O-])OC
IUPAC Name 1,2-dimethoxy-4,5-dinitrobenzene
Molecular Weight (g/mol) 228.16
PubChem CID 222989
Formula Weight 168.22
Percent Purity 97%
Chemical Name or Material N-(3-Hydroxyphenyl)thiourea
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'