Learn More
Macherey-Nagel™ N 20 PE Closures
Seal vials with polyethylene caps. Macherey-Nagel™ N 20 PE closues with no liner have a center hole and crimp neck.
104.00 NOK
Specifications
Color | Transparent |
---|---|
Material | Polyethylene |
Septum | No liner |
Quantity | 100 Pk. |
Thickness (Metric) | 1.3 mm |
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (Metric) | Price | Quantity & Availability | |||||
12480410
|
Macherey-Nagel™
70266 |
8.4 mm |
104.00 NOK
Pack of 100 |
Estimated Shipment: 18-04-2024
Log in to see stock. |
|||||
10192690
|
Macherey-Nagel™
70267 |
9.1 mm |
104.00 NOK
Pack of 100 |
Estimated Shipment: 24-04-2024
Log in to see stock. |
|||||
Description
- PE caps N 20 and suitable single septa N 19
- used as washer bottle closures
- Also used as interim closures
- PE caps N 20 and suitable single septa N 19
- Used as washer bottle closures
- Also used as interim closures
Specifications
Transparent | |
No liner | |
1.3 mm |
Polyethylene | |
100 Pk. | |
Crimp Cap |