missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Chloroethyl)pyrrolidine hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-3212915
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7250-67-1
C6H12ClN·HCl
170.08
MFCD00012718
FSNGFFWICFYWQC-UHFFFAOYSA-N
1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
81668
1-(2-chloroethyl)pyrrolidine;hydrochloride
C1CCN(C1)CCCl.Cl
This item is not returnable. View return policy
7250-67-1
C6H12ClN·HCl
170.08
MFCD00012718
FSNGFFWICFYWQC-UHFFFAOYSA-N
1-2-chloroethyl pyrrolidine hydrochloride, n-2-chloroethyl pyrrolidine hydrochloride, 2-chloroethylpyrrolidine hydrochloride, 2-pyrrolidinoethyl chloride hydrochloride, pyrrolidine, 1-2-chloroethyl-, hydrochloride, unii-yyq1h343r5, pyrrolidinoethyl chloride, hydrochloride, 1-2-chloroethyl pyrrolidine.hcl, 1-2-chloroethyl pyrrolidine hcl, 2-chloroethylpyrrolidinehydrochloride
81668
1-(2-chloroethyl)pyrrolidine;hydrochloride
C1CCN(C1)CCCl.Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 25 g
Beilstein 20, II, 4
Solubility Information Solubility in water: soluble
Formula Weight 170.08
Percent Purity 98%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material N-(2-Chloroethyl)pyrrolidine hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • N-(2-Chloroethyl)pyrrolidine hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'