Learn More
MYL4 Mouse anti-Human, Unconjugated, Clone: 2F5C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67533-1-IG-150UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16850216
  • 3828.53 NOK / 150µL
Estimated Shipment: 16-10-2023
to see stock.
Description
Description
The MYL4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MYL4. This antibody recognizes human, mouse, rat, pig antigen. The MYL4 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
MYL4
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat, Pig
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
2F5C1
Unconjugated
150 μL
4635
IgG2b