Learn More
MYH6 Monoclonal Antibody (CL2148), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA524631
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: QVEEDKVNSL SKSKVKLEQQ VDDLEGSLEQ EKKVRMDLER AKRKLEGDLK LTQESIMDLE NDKLQLEEKL KKKEFDINQQ NSKIEDEQVL ALQLQKKLKE NQARIEELEE ELEAERTARA KVEKLRSDLS RELEEISERL EEA Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 96%. Binds to an epitope located within the peptide sequence LTQESIMDLENDKLQ as determined by overlapping synthetic peptides.
MYH6 Monoclonal antibody specifically detects MYH6 in Human samples. It is validated for Immunohistochemistry, Western Blot.Specifications
MYH6 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P13533 | |
MYH6 | |
Recombinant protein corresponding to Human MYBPC2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry, Western Blot | |
CL2148 | |
Unconjugated | |
MYH6 | |
A830009F23Rik; AA517445; alpha cardiac MHC; alpha myosin; alpha-MHC; ASD3; cardiac myosin heavy chain alpha isoform; cardiomyopathy, hypertrophic 1; CMD1EE; CMH14; MYH6; MYHC; Myhca; Myhc-a; MyHC-alpha; myosin 6; Myosin heavy chain 6; myosin heavy chain polypeptide 6 cardiac muscle adult; myosin heavy chain, cardiac muscle alpha isoform; Myosin heavy chain, cardiac muscle alpha isoform (MyHC-alpha); myosin heavy chain, cardiac muscle, adult; myosin heavy chain, polypeptide 6, cardiac muscle, alpha; myosin, heavy chain 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha; myosin, heavy polypeptide 6, cardiac muscle, alpha (cardiomyopathy, hypertrophic 1); myosin-6; SSS3 | |
Mouse | |
Protein A | |
RUO | |
4624 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Research Use Only