Learn More
Thermo Scientific™ Multiskan Sky Microplate Spectrophotometer
Multiskan Sky Microplate Spectrophotometer is an absorbance microplate reader designed for the ultimate user experience. By introducing an attractive, touchscreen based user interface combined with multiple connectivity options, we offer refined photometry and bring unmatched versatility to academic, biotech, and pharmaceutical laboratories.
103700.00 NOK - 149400.00 NOK
Products 5
Product Code Brand Description Interface Price Quantity & Availability  
Product Code Brand Description Interface Price Quantity & Availability  
15797777
View Documents
Thermo Scientific™
51119500
Multiskan Sky SkanIt PC software
103700.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment
17-02-2021
Add to basket
15728147
View Documents
Thermo Scientific™
51119600
Multiskan Sky with touchscreen Touchscreen display and SkanIt PC software
121000.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment
17-02-2021
Add to basket
15738147
View Documents
Thermo Scientific™
51119600DP
Multiskan Sky with touchscreen and μDrop Plate Touchscreen display and SkanIt PC software
132200.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment
17-02-2021
Add to basket
15748147
View Documents
Thermo Scientific™
51119700
Multiskan Sky with touchscreen and cuvette Touchscreen display and SkanIt PC software
138300.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment
17-02-2021
Add to basket
15758147
View Documents
Thermo Scientific™
51119700DP
Multiskan Sky with touchscreen, cuvette and μDrop Plate Touchscreen display and SkanIt PC software
149400.00 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment
17-02-2021
Add to basket
Description
Description
  • Monochromator-based optical system for free selection of wavelengths from 200 nm to 1000 nm
  • Compatible with 96- and 384-well microplates with and without lids or cuvettes
  • Reads μDrop™ Plate for micro-volume DNA/RNA and protein analysis
  • Performs onboard shaking and incubation for temperature-critical or cell-based assays
  • Allows kinetic, spectral, and endpoint measurements for a variety of spectrophotometer applications
  • Fast operation: full spectrum of a well is done in less than 10 seconds and a full 96-well microplate is read in 6 seconds
  • Touchscreen models have an easy-to-use interface for stand-alone use and includes ready-made protocols for nucleic acid and protein quantification
  • Setting up of demanding assays is easy via the powerful and intuitive SkanIt™ Software
  • Thermo Fisher Connect cloud-based tools allow you to securely store, access, share, and manage your photometric data remotely (touchscreen models)
  • Multiple languages available on all models; choose between English, German, French, Spanish, Italian, Portuguese, Russian, Chinese, and Japanese
  • Automation compatibility for integration with SkanIt Software automation interface
SDS