missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi SMART Reagent Dispenser Accessories.

Brand:  Thermo Scientific™ N15136

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 15441825

  • 29696.00 NOK / Pack of 5


Estimated Shipment
21-01-2021
Add to basket

Description

Description

  • Cat. No. N15133 to N15139 are to be used with the Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
  • Cat. No. 24073292, 24073001, 24073297, 24073002, 24072675, 24072678 and 24072676 are to be used with the Multidrop Combi with SMART option instrument with serial numbers up to 836-2845SM
Specifications

Specifications

Autoclavable 10 times at 121°C, 1 bar, 20 min
Tubing: 0.4 mm; Tip 0.22 mm
Multidrop Combi with SMART2 option instrument with serial numbers 836-2846SM onwards
0.5 to 50 µL
SMART 2 Small tube metal tip dispensing cassette, new RFID tag, 5 pack
Stainless Steel Tip, Silicone Tubing
<1mL
Videos
SDS
Documents