Learn More
MSH2 Mouse anti-Human, Clone: 2H5F5, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 60161-1-IG-20UL
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16887135
  • 1281.92 NOK / 20µL
Estimated Shipment: 15-12-2022
Description
Plays a role in neurogenesis and neuronal migration By similarity. Necessary for correct formation of mitotic spindles and chromosome separation during mitosis. Necessary for cytokinesis and cell proliferation.
Specifications
Specifications
MSH2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P43246
MSH2
MSH2 Fusion Protein Ag7835
20 μL
Primary
Human
Antibody
IgG1
Western Blot, Immunohistochemistry (Paraffin)
2H5F5
Unconjugated
MSH2
COCA1, FCC1, hMSH2, HNPCC, HNPCC1, LCFS2, MSH2, MutS protein homolog 2
Mouse
Caprylic Acid Precipitation
RUO
4436
-20°C