missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum Labs™ mPES MicroKos™ Filter Modules
Spectrum Labs™ mPES MicroKos™ Filter Modules are single-use, hollow fiber filters designed for tangential flow filtration.
1455.00 NOK
Specifications
Material | Modified PES |
---|---|
Sterility | Non-sterile |
Includes | Inlet/Outlet Ports: Male Luer-Lok, Side Port: Female Luer-Lok |
Type | Hollow Fiber Filtration Module |
Diameter (Metric) | Housing OD: 4.6 mm |
Product Code | Brand | Length (Metric) | MWCO | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | MWCO | Price | Quantity & Availability | ||||
12457051
|
Spectrum Labs™
C02-E010-05-N |
Total Length: 23cm, Effective length: 20cm | 10 kDa |
1455.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
12477051
|
Spectrum Labs™
C02-E030-05-N |
Total Length: 23cm, Effective length: 20cm | 30 kDa |
1455.00 NOK
Each |
Estimated Shipment: 30-05-2024
Log in to see stock. |
||||
Description
- Designed for tangential flow filtration of small volumes ranging from 1mL to 100mL
- Single-use MicroKros™ hollow fiber filters are the first efficient and practical tangential flow filtration device for characterizing a process for scale-up or filtration of small volumes in a laboratory setting.
- MicroKros™ are available with MLL x FLL connections and 1 Effective Length
Specifications
Modified PES | |
Inlet/Outlet Ports: Male Luer-Lok, Side Port: Female Luer-Lok | |
Housing OD: 4.6 mm |
Non-sterile | |
Hollow Fiber Filtration Module | |
Fiber ID: 0.5mm, Housing ID: 3.1mm, Inlet/Retentake ID: 3mm, Side Port ID: 4.3mm |