Learn More
MP Biomedicals™ Linbro™ Screw-Cap Microcentrifuge Tubes
Suitable for researchers working with light sensitive samples. It protects precious light sensitive cells and cell lines.
Brand: MP Biomedicals™ 0215842490
Additional Details : Weight : 0.01000kg
Description
Screw Caps are not included. (Mfr. No. 0215843090)Specifications
1.5 mL | |
Plastic | |
Microcentrifuge tubes | |
Not Included |
Amber | |
Plastic amber tube | |
500 ea. | |
Cone |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.