Learn More
MP Biomedicals™ CellMaxx™ Bovine Albumin, Low IgG, Stem Cell Culture
Lipid-rich bovine serum albumin, suitable for stem cell culture and media formulation
Brand: MP Biomedicals™ 0219989790
Additional Details : CAS Number : 9048-46-8 Weight : 0.50000kg
Description
Chromatographically purified for culturing embryonic, mesenchymal and hematopoietic cells; IgG removed to very low levels. Lipid-rich formulation can be substituted for Albumax I™.
Physical characteristics:
- Form: Off white to brown powder
- Purity: ≥98% (w/w)
- Total Protein: ≥96%
- IgG 0.005% w/w
- pH (4%): 6.5 to 7.5
- Solubility: Dissolves in 20 minutes at 20° to 25°C
- Moisture: ≤5% w/w
- Endotoxin: ≤1EU/mg
- Mycoplasma: Negative
- Bioburden: ≤100cfu/g
- Virus (9CFR): Negative
- Reported: Lipid content
Specifications
ELISA, Mammalian Cell Culture, Western Blotting | |
Lipid-rich | |
CellMaxx™ Bovine Albumin, Low IgG | |
Chromatography | |
Bovine |
Powder | |
66 | |
6.5 to 7.5 | |
500g | |
4°C |