missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Albumin, Bovine, Fatty Acid-Free
Prepared by Fraction V method
POA
Specifications
For Use With (Application) | Immunohematology, Serology |
---|---|
Molecular Weight (g/mol) | 66.338 |
Name | Bovine Albumin |
pH Range | 5.2 to 7 |
Source | Bovine |
Description
- Low fatty acids
- Purity: >98%
- pH: 7
For best cell culture performance, see alternate Mfr. No. 180575
Specifications
Immunohematology, Serology | |
Bovine Albumin | |
Bovine | |
Powder |
66.338 | |
5.2 to 7 | |
2° to 8°C |