Learn More
ACROBiosystems Mouse Neuropilin-1 / NRP1 / CD304 Protein, Mouse IgG2a Fc Tag, low endotoxin
low endotoxin
Brand:  ACROBiosystems NR1-M5254-1mg
Additional Details : Weight : 0.00010kg
 View more versions of this product
Product Code. 16447929
  • 39223.36 NOK / 1mg
Estimated Shipment: 25-04-2024
to see stock.
Description
Description
Neuropilin-1 (NRP1) is also known as Vascular endothelial cell growth factor 165 receptor (VEGF165R), CD antigen CD304, which belongs to the neuropilin family. The membrane-bound isoform 1 is a receptor involved in the development of the cardiovascular system, in angiogenesis, in the formation of certain neuronal circuits and in organogenesis outside the nervous system. It mediates the chemorepulsant activity of semaphorins. It binds to semaphorin 3A, The PLGF-2 isoform of PGF, The VEGF-165 isoform of VEGF and VEGF-B. Coexpression with KDR results in increased VEGF-165 binding to KDR as well as increased chemotaxis. It may regulate VEGF-induced angiogenesis. The soluble isoform 2 binds VEGF-165 and appears to inhibit its binding to cells.
This protein carries a mouse IgG2a Fc tag at the C-terminus. The protein has a calculated MW of 120.5 kDa. The protein migrates as 125 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Specifications
Specifications
NP_032763.2
1 mg
Phe 22 - Pro 856
NRP1,Neuropilin-1,NRP,VEGF165R,CD304
Unconjugated
Mouse IgG Fc Tag
Less than 0.1 EU / μg by the LAL method.
Powder
>90% as determined by SDS-PAGE
120.5 kDa
Mouse NRP1, Mouse IgG2a Fc Tag, low endotoxin (NR1-M5254) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Pro 856 (Accession # P97333-1).
-20°C
Neuropilin-1
Mouse
50 mM Tris, 100 mM Glycine, pH7.5
HEK293
Bio-Markers and CD Antigens