missing translation for 'onlineSavingsMsg'
Learn More

Human IFN gamma, Recombinant Protein, Carrier-Free, Invitrogen™

Recombinant Protein

Brand:  Affymetrix eBioscience 34-8319-85

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15588776

  • 4825.00 NOK / 500µg


Estimated Shipment
08-09-2020
Add to basket

Description

Description

IFN gamma, also called Type II interferon, macrophage activation factor, and immune interferon is produced primarily by T-lymphocytes and natural killer cells in response to antigens, mitogens, Staphylococcus enterotoxin B, phytohemaglutanin and other cytokines. IFN gamma is a glycoprotein that exists, functionally, as a homodimer of approximately 45 kDa. On SDS-PAGE it appears as a combination of 25, 20 and minor 15.5kDa bands as a result of differential glycosylation. The biological activity of the IFN gamma homodimer is highly species specific and includes the following: antiviral activity, tumor antiproliferative activity, induction of class I and II MHC, macrophage activation, and enhanced immunoglobulin secretion by B lymphocytes. IFNg is involved in cytokine regulation and also acts synergistically with other cytokines. Description: Human IFN gamma is an ∽20kDa factor produced by activated T, B and NK cells and is an anti-viral and anti-parasitic cytokine. IFN gamma in synergy with other cytokines, such as TNF alpha, inhibits proliferation of normal and transformed cells. Immunomodulatory effects of IFN gamma are exerted on a wide range of cell types expressing the high affinity receptors for IFN-gamma. Glycosylation of IFN gamma does not affect its biological activity. Applications Reported: Recombinant human IFN gamma is biologically active and is for bioassay use only. It has been demonstrated to protect A549 cell line against viral infection in culture. For an ELISA standard, please refer to eBioscience Cat. No. 29-8319-65. Applications Tested: The ED50 of this protein, as measured by EMC virus protection assay using A549 cells, is less than or equal to 175 pg/mlL This corresponds to a specific activity of greater than or equal to 5.7 x 10e6 Units/mg. Source: E. coli expressed amino acids Gln24-Gln166 of mature human IFN gamma accession # NM_000619. Bioactivity: The ED50 of this protein, as measured by EMC virus protection assay using A549 cells, is less than or equal to 175 pg/mL. This corresponds to a specific activity of greater than or equal to 5.7 x 10e6 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: The methionylated N-terminal protein migrates as a 16kDa polypeptide on SDS-PAGE. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment.
Specifications

Specifications

P01579
Control, Functional Assay
Cytokine Bioassays
PBS with no preservative; pH 7.2
The methionylated N-terminal protein migrates as a 16kDa polypeptide on SDS-PAGE.
7.2
Affinity chromatography
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
IFNG
Recombinant Protein, Carrier-Free
Unlabeled
E. coli
Recombinant
0.5mg/mL
Liquid
3458
Human IFN gamma Carrier-Free
SDS-PAGE
500μg
-80°C
E. coli expressed amino acids Gln 24-Gln 166 of mature human IFN gamma accession # NM_000619
Human IFN gamma Carrier-Free
The ED50 of this protein, as measured by EMC virus protection assay using A549 cells, is less than or equal to 175pg/mL. This corresponds to a specific activity of greater than or equal to 5.7 x 106 Units/mg.
Human
Hu
Recombinant
Videos
Documents