missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse E-Cadherin Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB7482-SP
This item is not returnable.
View return policy
Description
E-Cadherin Monoclonal specifically detects E-Cadherin in Mouse samples. It is validated for ELISA Capture (Matched Antibody Pair).
Specifications
| E-Cadherin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Arc-1, CAD1, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CDH1, CDHE, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, LCAM, L-CAM, UVOE-Cadherin, uvomorulin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 999 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| ELISA | |
| 114403 | |
| ELISA Capture (Matched Antibody Pair) 2-8 ug/mL | |
| P09803 | |
| CDH1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse E-Cadherin Asp157-Val709 (predicted) Accession # P09803 | |
| 25 μg | |
| Primary | |
| Detects mouse E-Cadherin in ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant mouse (rm) VE-Cadherin, rmP-Cadherin, recombinant human (rh) Cadherin-17, or rhN-Cadherin is observed. | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction