missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Mouse CXCL10 (IP-10) Recombinant Protein
E. coli Recombinant Protein
1282.00 NOK - 2560.00 NOK
Specifications
Accession Number | P17515 |
---|---|
Concentration | 200 μg/mL |
For Use With (Application) | Control, ELISA, Western Blot |
Formulation | PBS with no preservative |
Gene ID (Entrez) | 15945 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17626695
|
Invitrogen™
RP87898 |
10 μg |
1282.00 NOK
10µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
17636695
|
Invitrogen™
RP87899 |
25 μg |
2560.00 NOK
25µg |
Estimated Shipment: 30-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
IP-10 (interferon-gamma induced protein 10 kD; in mouse, also called cytokine responsive gene 2, Crg-2, or mob-1) is a highly inducible, primary response gene that belongs to the CXC chemokine superfamily. It was first cloned in 1985. The biological functions of IP-10 are still unclear. Like Mig, IP-10 has no activity on neutrophils. Its functions include stimulation of monocytes, natural killer and T-cell migration, regulation of T-cell and bone marrow progenitor maturation, modulation of adhesion molecule expression as well as inhibition of angiogenesis. Mig and IP-10 share the same receptor, CXCR3.Specifications
P17515 | |
Control, ELISA, Western Blot | |
15945 | |
Mouse CXCL10 (aa 22-98) | |
-20°C | |
0.01 ng/ μg cytokine | |
CXCL10 | |
Unconjugated | |
Liquid |
200 μg/mL | |
PBS with no preservative | |
7.8 kD | |
E. coli | |
RUO | |
10 kDa interferon gamma-induced protein; 10 kDa interferon-gamma induced protein precursor; C Cmotif chemokine; C x C motif chemokine; C7; CC motif chemokine; CCmotif chemokine; chemokine (C-X-C motif) ligand 10; chemokine C-X-C motif ligand 10; chemokine CXC-like protein; Crg2; crg-2; CXC; CXC chemokine; CXC motif chemokine; C-X-C motif chemokine 10; C-X-C motif chemokine 10-like protein; C-X-C motif chemokine ligand 10; CXCL; CXCL10; CXCL10(1-73); gamma interferon inducible protein 10; gamma IP10; gamma-IP10; gIP-10; H-IP-10; IFI10; Inp10; interferon activated gene 10; interferon-gamma induced protein CRG-2; interferon-gamma-induced protein CRG-2; interferon-gamma-inducible protein 10; interferon-gamma-inducible protein-10; interferon-inducible cytokine IP-10; interferon-inducible protein 10; IP10; IP-10; M-IP-10; Mob1; mob-1; protein 10 from interferon (gamma)-induced cell line; protein Mob-1; Scyb10; similar to Small inducible cytokine B10 precursor (CXCL10) (10 kDa interferon-gamm | |
Mouse | |
Recombinant | |
>95% |