missing translation for 'onlineSavingsMsg'
Learn More

MOPS, Ultrapure Bioreagent, J.T. Baker™

Catalog Number p-5391236
21983.00 NOK valid until 2024-12-31
Use promo code "23268" to get your promotional price.


Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
mops, 4-morpholinepropanesulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-n-morpholinopropanesulfonic acid, unii-273bp63nv3
70807
CHEBI:44115
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
This item is not returnable. View return policy
1132-61-2
C7H15NO4S
209.26
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
mops, 4-morpholinepropanesulfonic acid, 3-morpholinopropanesulfonic acid, 3-n-morpholino propanesulfonic acid, 3-morpholinopropane-1-sulfonic acid, morpholinopropane sulfonic acid, morpholinopropanesulfonic acid, 4-morpholinepropane sulfonic acid, 3-n-morpholinopropanesulfonic acid, unii-273bp63nv3
70807
CHEBI:44115
[O-]S(=O)(=O)CCC[NH+]1CCOCC1

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

    For Molecular Biology Applications
Flash Point 116.00°C
Packaging Poly Bottle
Quantity 500 g
IUPAC Name 3-morpholin-4-ylpropane-1-sulfonic acid
Formula Weight 209.26
Grade Ultrapure Bioreagent
Chemical Name or Material MOPS
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'